Please login to your online account to display your discounted pricing

Calcium Chloride, Dihydrate, Calbiochem™, EMD Millipore

$107.26 - $107.26

Chemical Identifiers

Assay Percent Range ≥99%
Formula Weight 147
CAS 10035-04-8
Chemical Name or Material Calcium Chloride, Dihydrate
Molecular Formula CaCl2·2(H2O)
Purity Grade Notes by complexometry
Solubility Information Easily soluble in cold water
Recommended Storage Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).
Synonym CaCl2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

2082901KG

 
emd millipore corporation
2082901KG
Each for $107.26
Description & Specifications

Specifications

Formula Weight 147
Quantity 250g
CAS 10035-04-8
Assay Percent Range ≥99%
Synonym CaCl2
Purity Grade Notes by complexometry
Recommended Storage Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).
Chemical Name or Material Calcium Chloride, Dihydrate
Solubility Information Easily soluble in cold water
RTECS Number EV9810000
Purity Molecular Biology Grade
Molecular Formula CaCl2·2(H2O)

Hygroscopic

Safety and Handling

Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 20°C (68°F).