Please login to your online account to display your discounted pricing

Calcium Chloride Dihydrate, ACS Reagent Grade, Ricca Chemical


Quantity 25kg
Packaging Drum, Fiber

Manufacturer: ricca chemical company  RIRDCC007525F4

 View more versions of this product

Catalog No. RDCC007525F

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade ACS Reagent Grade
Physical Form Powder or Granules
Color White
Quantity 25kg
CAS Min % 98.0
CAS Max % 100.0
Chemical Name or Material Calcium Chloride Dihydrate
Packaging Drum, Fiber
CAS 10035-04-8

Specifications

Chemical Name or Material Calcium Chloride Dihydrate
CAS 10035-04-8
CAS Min % 98.0
CAS Max % 100.0
Purity Grade ACS Reagent Grade
Physical Form Powder or Granules