Please login to your online account to display your discounted pricing

Calcium Chloride Dihydrate, ACS Reagent Grade, Ricca Chemical

$1,639.88 - $1,639.88

Chemical Identifiers

Chemical Name or Material Calcium Chloride Dihydrate
CAS 10035-04-8
CAS Min % 98.0
CAS Max % 100.0
Purity Grade ACS Reagent Grade
Physical Form Powder or Granules
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

RDCC007525F

 
ricca chemical company
RIRDCC007525F4
Drum, Fiber 25kg Each for $1,639.88

RDCC0075500

 
ricca chemical company
RIRDCC0075500B1
Poly Wide mouth 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder or Granules
Purity Grade ACS Reagent Grade
Color White
Quantity 25kg
CAS Min % 98.0
CAS Max % 100.0
Chemical Name or Material Calcium Chloride Dihydrate
Packaging Drum, Fiber
CAS 10035-04-8