Please login to your online account to display your discounted pricing

Calcium Chloride, Dihydrate, ACS Grade, 99.0 to 105.0%, LabChem™

$141.69 - $141.69

Chemical Identifiers

Chemical Name or Material Calcium Chloride
CAS 10035-04-8
CAS Max % 100
Name Note Dihydrate
Purity Grade ACS Grade
Linear Formula CaCl2·2H2O
Formula Weight 147.02
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC127251

 
lab chem
LC127251
500g Each for $141.69
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 147.02
Chemical Name or Material Calcium Chloride
EINECS Number 233-140-8
Identification Passes Test
Name Note Dihydrate
Packaging Poly Bottle
Purity Grade ACS Grade
Assay 99.0 to 105.0%
Solubility Soluble in water
Specific Gravity 0.835g/cm3
Storage Information Room Temperature
Linear Formula CaCl2·2H2O
CAS 10035-04-8
Decomposition Information Hydrogen chloride
Color White
Quantity 500g
Health Hazard 1 Warning
CAS Max % 100
Health Hazard 2 GHS H Statement
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Density 0.835g/cm3
Safety and Handling

Warning

GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
Causes serious eye irritation.