Please login to your online account to display your discounted pricing

Calcium Chloride, Dihydrate, ACS, 99.0 to 102.0%, EMD Millipore

$114.86 - $622.63

Chemical Identifiers

Chemical Name or Material Calcium Chloride, Dihydrate
Name Note EMSURE™, For Analysis
CAS 10035-04-8
Purity Grade ACS/EP/Reagent Grade
Physical Form Powder
Assay Percent Range 99.0 to 102.0%
Molecular Formula CaCl2·H2O
Linear Formula CaCl2·H2O
Formula Weight 147.2
Synonym Calcium dichloride
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1023820500

 
emd millipore corporation
1023820500
Plastic Bottle 500g Each for $114.86

M1023825000

 
emd millipore corporation
1023825000
Plastic Bottle 5kg Each for $622.63
Description & Specifications

Specifications

Purity Grade ACS/EP/Reagent Grade
Physical Form Powder
Formula Weight 147.2
Color White
Quantity 500g
Density 1.85g/cm3 (2°C)
Chemical Name or Material Calcium Chloride, Dihydrate
Name Note EMSURE™, For Analysis
Packaging Plastic Bottle
Assay Percent Range 99.0 to 102.0%
Molecular Formula CaCl2·H2O
Linear Formula CaCl2·H2O
CAS 10035-04-8
Synonym Calcium dichloride
Recommended Storage Store without Restrictions
Safety and Handling

Store without Restrictions