Please login to your online account to display your discounted pricing

Calcium Chloride Dihydrate, ∽99%, MP Biomedicals


Quantity 500g

Manufacturer: mp biomedicals  0219508890

 View more versions of this product

Catalog No. ICN19508890

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 500g
Chemical Name or Material Calcium Chloride Dihydrate
CAS 10035-04-8
Assay Percent Range ∽99%
Molecular Formula CaCl2 · 2H2O
Formula Weight 147g/mol
Recommended Storage Room Temperature

Specifications

Chemical Name or Material Calcium Chloride Dihydrate
Assay Percent Range ∽99%
Molecular Formula CaCl2 · 2H2O
Formula Weight 147g/mol
Accessories

Room Temperature