missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium chloride dihydrate, 99% min., Alfa Aesar™

Manufacturer:  Alfa Aesar 1231236

 View more versions of this product

Catalog No. AA1231236



Specifications

Specifications

Calcium chloride dihydrate
99% min.
MFCD00149613
Granular
500g
10035-04-8
CaCl2·2H2O
147.02 (110.99anhydrous)
0.835g/mL

Chemical Identifiers

10035-04-8
CaCl2·2H2O
MFCD00149613
Safety and Handling

Safety and Handling

EINECSNumber : 233-140-8

Documents