missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium chloride dihydrate, 99+%, for analysis, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 207780010

 View more versions of this product

Catalog No. AC207780010



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent
Specifications

Specifications

Calcium chloride dihydrate
White
4.5 to 6.5 (5% soln. at 20°C)
1 kg
CaCl2·2H2O
01,104
Solubility in water: 1000g/L (0°C). Other solubilities: freely soluble in alcohol
147.02
Analytical
0.83
175.0°C
10035-04-8
Crystalline Powder and/or Crystals
99% min. (Complexometry)
CaCl2·2H2O
15,1661
147.02
≥99%
Plastic Bottle
0.8300g/mL

Chemical Identifiers

10035-04-8
CaCl2·2H2O
147.02
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only