Please login to your online account to display your discounted pricing

Calcium Chloride, Certified, 0.0250M ±0.0003M (0.05N), LabChem™

$50.37 - $50.37

Chemical Identifiers

Chemical Name or Material Calcium Chloride
CAS 7732-18-5
CAS Max % 99.63
CAS 10035-04-8
CAS Max % 0.37
Purity Grade Certified Grade
Linear Formula CaCl2·2H2O
Formula Weight 147.02
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC128051

 
lab chem
LC128051
500mL Each for $50.37
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 147.02
Physical Form Liquid
Decomposition Information Hydrogen chloride
Linear Formula CaCl2·2H2O
Traceability to NIST Traceable to NIST
CAS 7732-18-5
Quantity 500mL
Identification Passes Test
Concentration 0.0250M ±0.0003M (0.05N)
Packaging Poly Bottle
Color Clear, colorless
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
CAS Max % 0.37
Storage Information Room Temperature
CAS 10035-04-8
Chemical Name or Material Calcium Chloride
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Solubility Soluble in water
CAS Max % 99.63
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.