Please login to your online account to display your discounted pricing

Calcium Chloride, Certified, 0.0200M ±0.0002M (0.04N), LabChem™

$41.71 - $41.71

Chemical Identifiers

Chemical Name or Material Calcium Chloride
CAS 7732-18-5
CAS Max % 99.71
CAS 10035-04-8
CAS Max % 0.29
Purity Grade Certified Grade
Linear Formula CaCl2·2H2O
Formula Weight 147.02
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC128001

 
lab chem
LC128001
500mL Each for $41.71
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 147.02
CAS Max % 99.71
CAS 10035-04-8
CAS 7732-18-5
Physical Form Liquid
Linear Formula CaCl2·2H2O
Storage Information Room Temperature
Traceability to NIST Traceable to NIST
Identification Passes Test
Color Clear, colorless
Chemical Name or Material Calcium Chloride
Solubility Soluble in water
Decomposition Information Hydrogen chloride
Quantity 500mL
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
CAS Max % 0.29
Concentration 0.0200M ±0.0002M (0.04N)
Packaging Poly Bottle
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.