missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Calcium chloride, anhydrous, 93%, Alfa Aesar™
Manufacturer: Alfa Aesar 12316A1
Specifications
Calcium chloride | |
93% | |
MFCD00010903 | |
calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal | |
[Cl-].[Cl-].[Ca+2] | |
110.978 | |
CHEBI:3312 | |
Granular Powder | |
White to Gray |
10043-52-4 | |
CaCl2 | |
White to gray granular powder | |
UXVMQQNJUSDDNG-UHFFFAOYSA-L | |
calcium;dichloride | |
5284359 | |
110.99 | |
3.08g/cm3 | |
1kg |
Chemical Identifiers
10043-52-4 | |
110.978 | |
UXVMQQNJUSDDNG-UHFFFAOYSA-L | |
5284359 | |
calcium;dichloride |
CaCl2 | |
MFCD00010903 | |
calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal | |
CHEBI:3312 | |
[Cl-].[Cl-].[Ca+2] |
Safety and Handling
H319
EINECSNumber : 233-140-8