Please login to your online account to display your discounted pricing

Calcium Chloride, Anhydrous, 20 Mesh and finer, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$416.07 - $1,592.10

Chemical Identifiers

Chemical Name or Material Calcium Chloride
CAS 10043-52-4
Name Note Anhydrous
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula CaCl2
Formula Weight 110.98
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002510

 
avantor performance
131105
Poly Bottle 2.5kg Each for $1,592.10

02002528

 
avantor performance
131101
Poly Bottle 500g Each for $416.07
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 110.98
Chemical Name or Material Calcium Chloride
Molecular Formula CaCl2
CAS 10043-52-4
Mesh Size 20 Mesh and finer
Name Note Anhydrous
Packaging Poly Bottle
Specific Gravity 1L = 2.15kg
Quantity 2.5kg

    For desiccant use