missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium Chloride ACS AR Powder, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 10035-04-8
Physical Form Powder
Molecular Formula CaCl2·2H2O
Formula Weight 147.01
Chemical Name or Material Calcium Chloride, Dihydrate
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK4160212 Avantor Performance
416006
Poly 2.5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

10035-04-8
CaCl2·2H2O
Calcium Chloride, Dihydrate
Powder
147.01
ACS Grade

Specifications

147.01
Poly
Powder
Calcium Chloride, Dihydrate
2.5kg
CaCl2·2H2O
10035-04-8
ACS Grade