Please login to your online account to display your discounted pricing

Calcium Chloride ACS AR Powder, Macron Fine Chemicals™


macron

Quantity 2.5kg
Packaging Poly

Manufacturer: avantor performance  416006

 View more versions of this product

Catalog No. MK4160212

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 147.01
Purity Grade ACS Grade
Quantity 2.5kg
Chemical Name or Material Calcium Chloride
Packaging Poly
Molecular Formula CaCl2·2H2O
Physical Form Powder
Name Note Dihydrate
CAS 10035-04-8

Specifications

Chemical Name or Material Calcium Chloride
Name Note Dihydrate
CAS 10035-04-8
Purity Grade ACS Grade
Physical Form Powder
Molecular Formula CaCl2·2H2O
Formula Weight 147.01