Please login to your online account to display your discounted pricing

Calcium Chloride ACS AR Powder, Macron Fine Chemicals™


macron

Quantity 500g
Packaging POLYSTORMOR

Manufacturer: avantor performance  416012

 View more versions of this product

Catalog No. MK4160500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Packaging POLYSTORMOR
Quantity 500g
Chemical Name or Material Calcium Chloride
Name Note Dihydrate
CAS 10035-04-8
Purity Grade ACS
Physical Form Powder
Molecular Formula CaCl2·2H2O
Formula Weight 147.01g/mol

Specifications

Chemical Name or Material Calcium Chloride
Name Note Dihydrate
CAS 10035-04-8
Purity Grade ACS
Physical Form Powder
Molecular Formula CaCl2·2H2O
Formula Weight 147.01g/mol