Please login to your online account to display your discounted pricing

Calcium chloride, 96%, for analysis, granules, ACROS Organics™


Quantity 500g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC219175000

 View more versions of this product

Catalog No. AC219175000

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 110.98
Quantity 500g
Fieser 01,104; 04,368
Merck Index 15, 1661
Purity Grade For analysis
Physical Form Granules
Chemical Name or Material Calcium chloride
Assay Percent Range 96%
Molecular Formula CaCl2
Linear Formula CaCl2
MDL Number MFCD00010903
CAS 10043-52-4
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note Granules
Packaging Plastic bottle

Specifications

Chemical Name or Material Calcium chloride
Name Note Granules
CAS 10043-52-4
Purity Grade For analysis
Physical Form Granules
Assay Percent Range 96%
Molecular Formula CaCl2
Linear Formula CaCl2
Formula Weight 110.98
MDL Number MFCD00010903
Accessories

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.

SDS & Safety Documents