missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Calcium chloride, 96%, for biochemistry, anhydrous, Thermo Scientific Chemicals
$464.08 - $464.08
Chemical Identifiers
CAS | 10043-52-4 |
---|---|
Molecular Formula | CaCl2 |
Molecular Weight (g/mol) | 110.98 |
InChI Key | UXVMQQNJUSDDNG-UHFFFAOYSA-L |
Synonym | calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal |
PubChem CID | 5284359 |
ChEBI | CHEBI:3312 |
IUPAC Name | calcium dichloride |
SMILES | [Cl-].[Cl-].[Ca++] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC192735000
|
Thermo Scientific Chemicals
192735000 |
500 g |
Each for $464.08
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10043-52-4 | |
110.98 | |
calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal | |
CHEBI:3312 | |
[Cl-].[Cl-].[Ca++] |
CaCl2 | |
UXVMQQNJUSDDNG-UHFFFAOYSA-L | |
5284359 | |
calcium dichloride |
Specifications
>1600.0°C | |
10043-52-4 | |
95% min. (Argentometry) | |
CaCl2 | |
15,1661 | |
Solubility in water: 740g/L (20°C). Other solubilities: freely soluble in alcohol | |
[Cl-].[Cl-].[Ca++] | |
110.98 | |
CHEBI:3312 | |
96% | |
Calcium chloride |
782.0°C | |
500 g | |
CaCl2 | |
01,104; 04,368 | |
calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal | |
UXVMQQNJUSDDNG-UHFFFAOYSA-L | |
calcium dichloride | |
5284359 | |
110.99 | |
Biochemical |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 233-140-8
RUO – Research Use Only