missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium chloride, 100mM aq. soln., sterile

$85.73 - $147.12

Chemical Identifiers

CAS 10043-52-4
Molecular Formula CaCl2
Molecular Weight (g/mol) 110.978
MDL Number MFCD00010903
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
Synonym calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
PubChem CID 5284359
ChEBI CHEBI:3312
IUPAC Name calcium;dichloride
SMILES [Cl-].[Cl-].[Ca+2]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62905AP View Documents Alfa Aesar™
J62905AP
500mL Each for $85.73
 
AAJ62905K2 View Documents Alfa Aesar™
J62905K2
1L Each for $147.12
 
Specifications

Chemical Identifiers

10043-52-4
110.978
UXVMQQNJUSDDNG-UHFFFAOYSA-L
5284359
calcium;dichloride
CaCl2
MFCD00010903
calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
CHEBI:3312
[Cl-].[Cl-].[Ca+2]

Specifications

10043-52-4
MFCD00010903
UXVMQQNJUSDDNG-UHFFFAOYSA-L
calcium;dichloride
5284359
110.99
100mM aq. soln.
Calcium chloride, sterile
CaCl2
calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
[Cl-].[Cl-].[Ca+2]
110.978
CHEBI:3312
Liquid
500mL
Safety and Handling

Safety and Handling

TSCA : TSCA

Recommended Storage : 2° to 8°C