missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium carbonate, Puratronic™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 4723506

 View more versions of this product

Catalog No. AA4723506



Specifications

Specifications

Calcium carbonate
Puratronic™
CCaO3
14,1657
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium;carbonate
10112
100.09
99.999%
Odorless
800°C (decomposition)
471-34-1
(metals basis)
MFCD00010906
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
C(=O)([O-])[O-].[Ca+2]
100.086
CHEBI:3311
Powder
2.930g/mL
White
5g

Chemical Identifiers

471-34-1
100.086
VTYYLEPIZMXCLO-UHFFFAOYSA-L
10112
calcium;carbonate
CCaO3
MFCD00010906
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
CHEBI:3311
C(=O)([O-])[O-].[Ca+2]
Safety and Handling

Safety and Handling

RTECSNumber : FF9335000

TSCA : TSCA

SDS Documents