Learn More
Calcium carbonate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04723514
Description
Applied as neutralizing agent, filler, and extender in rubber, plastics, paints, opacifying agent in paper, putty, antacid, Portland cement, sulfur dioxide, removal from stack gases, allurgical flux, analytical chemistry, carbon dioxide generation, and determining halogens in organic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Calcium carbonate | |
471-34-1 | |
Powder | |
(metals basis) | |
MFCD00010906 | |
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama | |
VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
calcium carbonate | |
10112 | |
100.09 | |
Puratronic™ | |
2.930 g/mL |
800°C (decomposition) | |
White | |
25 g | |
CCaO3 | |
14,1657 | |
Soluble in dilute acids (5 M HCl: 0.1 M at 20°C,clear,colorless). Insoluble in water. | |
[Ca++].[O-]C([O-])=O | |
100.09 | |
CHEBI:3311 | |
99.999% | |
Odorless |
Chemical Identifiers
471-34-1 | |
100.09 | |
VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
10112 | |
[Ca++].[O-]C([O-])=O |
CCaO3 | |
MFCD00010906 | |
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama | |
CHEBI:3311 |
Safety and Handling
EINECSNumber : 207-439-9
RTECSNumber : FF9335000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only