Learn More
Calcium carbonate, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01067906
Description
Calcium carbonate is used in medicine and agriculture. It is also used in flue gas desulfurization in fossil fuel power stations, production of toothpaste, ceramics/glazing applications, purification of iron from iron ore, and to clean tarnish on silver. It forms the major component of ceramic tile adhesives. It is used in the refining of sugar, as filler in thermosetting resins, as an abrasive, as an extender in paints, and as a formation-bridging filtercake-sealing agent in oil industry. It is used in swimming pools for controlling the pH, to improve the rigidity of polypropylene compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Calcium carbonate | |
471-34-1 | |
Powder | |
(metals basis) | |
MFCD00010906 | |
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama | |
VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
calcium;carbonate | |
10112 | |
100.09 | |
Puratronic™ | |
1.6583 |
800°C (decomposition) | |
White | |
5 g | |
CCaO3 | |
14,1657 | |
Very low solubility in water. | |
[Ca++].[O-]C([O-])=O | |
100.09 | |
CHEBI:3311 | |
99.99% | |
Odorless | |
2.930 g/mL |
Chemical Identifiers
471-34-1 | |
100.09 | |
VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
10112 | |
calcium;carbonate |
CCaO3 | |
MFCD00010906 | |
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama | |
CHEBI:3311 | |
[Ca++].[O-]C([O-])=O |
Safety and Handling
EINECSNumber : 207-439-9
RTECSNumber : FF9335000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only