Please login to your online account to display your discounted pricing

Calcium Carbonate, Powder, Low in Alkalies, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$331.36 - $970.37

Chemical Identifiers

Chemical Name or Material Calcium Carbonate
CAS 471-34-1
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula CaCO3
Formula Weight 100.09
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002515

 
avantor performance
129405
Poly Bottle 2kg Each for $970.37

02002517

 
avantor performance
129401
Poly Bottle 500g Each for $331.36
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 100.09
Chemical Name or Material Calcium Carbonate
CAS 471-34-1
Molecular Formula CaCO3
Physical Form Powder
Packaging Poly Bottle
Quantity 2kg