missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium carbonate, 98+%, pure, heavy powder, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 124670010

 View more versions of this product

Catalog No. AC124670010



Description

Description

Specifications

Specifications

Calcium carbonate
98% min. (Complexometry)
CaCO3
15,1659
Solubility in water: insoluble. Other solubilities: soluble in diluted acids
C(=O)([O-])[O-].[Ca+2]
100.09
CHEBI:3311
Powder
Pure
Solubility in water: insoluble. Other solubilities: soluble in diluted acids
825.0°C
471-34-1
CCaO3
01,103; 02,57; 04,67; 05,89
aeromatt, akadama, aragonite, calcite, calcium carbonate, calofort u, carbonic acid calcium salt 1:1, chalk, limestone, marble
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium;carbonate
10112
100.09
98+%
Plastic bottle
White
1kg

Chemical Identifiers

471-34-1
100.09
aeromatt, akadama, aragonite, calcite, calcium carbonate, calofort u, carbonic acid calcium salt 1:1, chalk, limestone, marble
CHEBI:3311
C(=O)([O-])[O-].[Ca+2]
CCaO3
VTYYLEPIZMXCLO-UHFFFAOYSA-L
10112
calcium;carbonate
Safety and Handling

Safety and Handling

EINECSNumber : 207-439-9

SDS Documents