Please login to your online account to display your discounted pricing

Calcium Carbonate, ACS Grade, ≥ 99.0%, LabChem™

$212.89 - $212.89

Chemical Identifiers

Chemical Name or Material Calcium Carbonate
CAS 471-34-1
CAS Max % 100
Purity Grade ACS Grade
Linear Formula CaCO3
Formula Weight 100.09
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC126901

 
lab chem
LC126901
500g Each for $212.89
Description & Specifications

Specifications

Formula Weight 100.09
Assay ≥99.0%
Color White
Packaging Poly Bottle
Melting Point 520°C
Chemical Name or Material Calcium Carbonate
Boiling Point 825°C
Quantity 500g
Physical Form Powder
CAS 471-34-1
Linear Formula CaCO3
Health Hazard 1 Warning
Specific Gravity 2.96g/cm3
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes eye irritation.
May cause respiratory irritation.
Storage Information Room Temperature
Density 2.96g/cm3
Identification Passes Test
CAS Max % 100
Purity Grade ACS Grade
Health Hazard 3 GHS P Statement
Avoid breathing dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store in a well-ventilated place.
Keep container tightly closed.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
EINECS Number 207-439-9
Solubility Soluble in water
Safety and Handling

Warning

GHS P Statement
Avoid breathing dust.
Use only outdoors or in a well-ventilated area.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store in a well-ventilated place.
Keep container tightly closed.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes skin irritation.
Causes eye irritation.
May cause respiratory irritation.