missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium Carbonate ACS AR Powder, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 471-34-1
Physical Form Powder
Molecular Formula CaCO3
Formula Weight 100.09
Chemical Name or Material Calcium Carbonate, Chelometric Standard
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK4071212 Avantor Performance
407105
Lined Fiber Dr 2.5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MK4071500 Avantor Performance
407112
Glass 500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

471-34-1
CaCO3
Calcium Carbonate, Chelometric Standard
Powder
100.09
ACS Grade

Specifications

471-34-1
CaCO3
Lined Fiber Dr
Calcium Carbonate, Chelometric Standard
Powder
100.09
2.5kg
ACS Grade