Please login to your online account to display your discounted pricing

Calcium Carbonate ACS AR Powder, Macron Fine Chemicals™


macron

Quantity 500g
Packaging Glass

Manufacturer: avantor performance  407112

 View more versions of this product

Catalog No. MK4071500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 100.09
Purity Grade ACS Grade
Physical Form Powder
Packaging Glass
Molecular Formula CaCO3
Name Note Chelometric Standard
Chemical Name or Material Calcium Carbonate
CAS 471-34-1
Quantity 500g

Specifications

Chemical Name or Material Calcium Carbonate
Name Note Chelometric Standard
CAS 471-34-1
Purity Grade ACS Grade
Physical Form Powder
Molecular Formula CaCO3
Formula Weight 100.09