Please login to your online account to display your discounted pricing

Calcium Carbonate, >99%, (Crystalline/ACS Reagent Grade), MP Biomedicals


Quantity 1kg

Manufacturer: mp biomedicals  0219141391

 View more versions of this product

Catalog No. ICN19141391

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1kg
Chemical Name or Material Calcium Carbonate
CAS 471-34-1
Purity Grade ACS Reagent
Physical Form Crystalline
Assay Percent Range >99%
Molecular Formula CaCO3
Formula Weight 100.1g/mol
Recommended Storage Room Temperature

Specifications

Chemical Name or Material Calcium Carbonate
CAS 471-34-1
Purity Grade ACS Reagent
Physical Form Crystalline
Assay Percent Range >99%
Molecular Formula CaCO3
Formula Weight 100.1g/mol
Accessories

Room Temperature