Please login to your online account to display your discounted pricing

Calcium Carbonate, >99%, (Crystalline/ACS Reagent Grade), MP Biomedicals

$74.42 - $420.65

Chemical Identifiers

Chemical Name or Material Calcium Carbonate
CAS 471-34-1
Purity Grade ACS Reagent
Physical Form Crystalline
Assay Percent Range >99%
Molecular Formula CaCO3
Formula Weight 100.1g/mol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN19141380

 
mp biomedicals
0219141380
100g Each for $74.42

ICN19141391

 
mp biomedicals
0219141391
1kg Each for $420.65
Description & Specifications

Specifications

Quantity 100g
Chemical Name or Material Calcium Carbonate
CAS 471-34-1
Purity Grade ACS Reagent
Physical Form Crystalline
Assay Percent Range >99%
Molecular Formula CaCO3
Formula Weight 100.1g/mol
Recommended Storage Room Temperature
Safety and Handling

Room Temperature