missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium carbonate, 99+%, for biochemistry, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC192721000

 View more versions of this product

Catalog No. AC192721000



Specifications

Specifications

Calcium carbonate
99% min. (Complexometry)
CaCO3
15,1659
Solubility in water: insoluble. Other solubilities: soluble in diluted acids
C(=O)([O-])[O-].[Ca+2]
100.09
CHEBI:3311
99+%
0.005% max
Plastic bottle
0.02% max
100g
471-34-1
CCaO3
01,103; 02,57; 04,67; 05,89
aeromatt
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium;carbonate
10112
100.09
Biochemistry
0.01% max. (in HCl)
Solubility in water: insoluble. Other solubilities: soluble in diluted acids
825.0°C

Chemical Identifiers

471-34-1
100.09
aeromatt
CHEBI:3311
C(=O)([O-])[O-].[Ca+2]
CCaO3
VTYYLEPIZMXCLO-UHFFFAOYSA-L
10112
calcium;carbonate
Safety and Handling

Safety and Handling

EINECSNumber : 207-439-9

SDS Documents