Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium carbonate, 99.95% (metals basis)

Chemical Identifiers

Chemical Name or Material Calcium carbonate
CAS 471-34-1
Formula Weight 100.09
MDL Number MFCD00010906
Merck Index 14,1657
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1099614

  SDS
alfa aesar™
1099614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1099622

  SDS
alfa aesar™
1099622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 100.09
CAS 471-34-1
Assay 99.95%
RTECS Number FF9335000
Assay Percent Notes (metals basis)
Chemical Name or Material Calcium carbonate
MDL Number MFCD00010906
Melting Point 800°C (dec.)
Refractive Index 1.6583
Density 2.930g/mL
Merck Index 14,1657
Physical Form Powder
Odor Odorless
Quantity 25g
TSCA TSCA