missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium carbonate, 99.95% (metals basis)

Manufacturer:  Alfa Aesar™ 1099622

 View more versions of this product

Catalog No. AA1099622



Specifications

Specifications

471-34-1
CCaO3
14,1657
Very low solubility in water.
C(=O)([O-])[O-].[Ca+2]
100.086
CHEBI:3311
Powder
2.930 g/mL
1.6583
100g
(metals basis)
MFCD00010906
aeromatt, akadama, aragonite, calcite, calcium carbonate, calofort u, carbonic acid calcium salt 1:1, chalk, limestone, marble
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium;carbonate
10112
100.09
99.95%
Odorless
800°C (decomposition)

Chemical Identifiers

471-34-1
100.086
VTYYLEPIZMXCLO-UHFFFAOYSA-L
10112
calcium;carbonate
CCaO3
MFCD00010906
aeromatt, akadama, aragonite, calcite, calcium carbonate, calofort u, carbonic acid calcium salt 1:1, chalk, limestone, marble
CHEBI:3311
C(=O)([O-])[O-].[Ca+2]
Safety and Handling

Safety and Handling

EINECSNumber : 207-439-9

RTECSNumber : FF9335000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents