Please login to your online account to display your discounted pricing

Calcium carbonate, 99.5% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Calcium carbonate
CAS 471-34-1
Molecular Formula CaCO3
Formula Weight 100.09
MDL Number MFCD00010906
Merck Index 14,1657
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA1140322

  SDS
alfa aesar
1140322
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 100.09
Quantity 100g
Chemical Name or Material Calcium carbonate
TSCA TSCA
Odor Odorless
Physical Form 5μm Powder
Refractive Index 1.6583
Assay 99.5%
Molecular Formula CaCO3
MDL Number MFCD00010906
CAS 471-34-1
Merck Index 14,1657
Color White
Melting Point 800°C (dec.)
Density 2.930
RTECS Number FF9335000