missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium carbonate, 99.5% (metals basis) , Alfa Aesar™

Manufacturer:  Alfa Aesar 11403A3

 View more versions of this product

Catalog No. AA11403A3



Specifications

Specifications

Calcium carbonate
99.5% (Metal basis)
MFCD00010906
aeromatt
C(=O)([O-])[O-].[Ca+2]
100.086
CHEBI:3311
Powder
2kg
471-34-1
CCaO3
White powder
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium;carbonate
10112
100.09
White

Chemical Identifiers

471-34-1
100.086
VTYYLEPIZMXCLO-UHFFFAOYSA-L
10112
calcium;carbonate
CCaO3
MFCD00010906
aeromatt
CHEBI:3311
C(=O)([O-])[O-].[Ca+2]
Safety and Handling

Safety and Handling

EINECSNumber : 207-439-9

Documents