missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium carbonate, 99.5% (metals basis) , Alfa Aesar™

Manufacturer:  Alfa Aesar 1140322

 View more versions of this product

Catalog No. AA1140322



Specifications

Specifications

Calcium carbonate
CCaO3
14,1657
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium;carbonate
10112
100.09
99.5%
Odorless
White
100g
471-34-1
MFCD00010906
aeromatt
C(=O)([O-])[O-].[Ca+2]
100.086
CHEBI:3311
5μm Powder
2.930
1.6583
800°C (decomposition)

Chemical Identifiers

471-34-1
100.086
VTYYLEPIZMXCLO-UHFFFAOYSA-L
10112
calcium;carbonate
CCaO3
MFCD00010906
aeromatt
CHEBI:3311
C(=O)([O-])[O-].[Ca+2]
Safety and Handling

Safety and Handling

RTECSNumber : FF9335000

TSCA : TSCA

Documents