Learn More
Calcium carbonate, 98%, pure, light powder, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 403815000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Calcium carbonate | |
98% | |
White | |
500 g | |
CCaO3 | |
MFCD00010906 | |
15, 1659 | |
Solubility in water: insoluble. Other solubilities: soluble in diluted acids | |
[Ca++].[O-]C([O-])=O | |
100.09 | |
CHEBI:3311 | |
98% | |
Plastic bottle |
825.0°C | |
471-34-1 | |
Fine Powder | |
97.5% min. (Complexometry) | |
CaCO3 | |
01,103; 02,57; 04,67; 05,89 | |
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama | |
VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
calcium;carbonate | |
10112 | |
100.09 | |
Pure |
Chemical Identifiers
471-34-1 | |
100.09 | |
VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
10112 | |
[Ca++].[O-]C([O-])=O |
CCaO3 | |
MFCD00010906 | |
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama | |
CHEBI:3311 |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
HYGROSCOPIC
EINECSNumber : 207-439-9
RTECSNumber : FF9335000
TSCA : TSCA
RUO – Research Use Only