missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Calcium carbonate, 98%, Alfa Aesar™
Manufacturer: Alfa Aesar 12365A3
Description
- Manufacture of paint, rubber, plastics, etc
- In food, cosmetics, and pharmaceuticals
- As soil conditioner, neutralizer of surface waters, and as an industrial acid neutralizer
Specifications
Calcium carbonate | |
98% | |
MFCD00010906 | |
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama | |
VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
calcium;carbonate | |
10112 | |
100.09 | |
98% | |
2kg |
471-34-1 | |
CCaO3 | |
14,1657 | |
Practically insoluble in water; Soluble in diluted acids | |
C(=O)([O-])[O-].[Ca+2] | |
100.086 | |
CHEBI:3311 | |
Powder | |
2.93g/mL |
Chemical Identifiers
471-34-1 | |
100.086 | |
VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
10112 | |
calcium;carbonate |
CCaO3 | |
MFCD00010906 | |
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama | |
CHEBI:3311 | |
C(=O)([O-])[O-].[Ca+2] |
Safety and Handling
RTECSNumber : FF9335000
TSCA : TSCA