missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium carbonate, 98%, Alfa Aesar™

Manufacturer:  Alfa Aesar 1236536

 View more versions of this product

Catalog No. AA1236536



Description

Description

  • Manufacture of paint, rubber, plastics, etc
  • In food, cosmetics, and pharmaceuticals
  • As soil conditioner, neutralizer of surface waters, and as an industrial acid neutralizer
Specifications

Specifications

Calcium carbonate
98%
MFCD00010906
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium;carbonate
10112
100.09
98%
Odorless
White
500g
471-34-1
CCaO3
14,1657
Practically insoluble in water; Soluble in diluted acids
C(=O)([O-])[O-].[Ca+2]
100.086
CHEBI:3311
Powder
2.930g/mL
1.6583
800°C (decomposition)

Chemical Identifiers

471-34-1
100.086
VTYYLEPIZMXCLO-UHFFFAOYSA-L
10112
calcium;carbonate
CCaO3
MFCD00010906
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
CHEBI:3311
C(=O)([O-])[O-].[Ca+2]
Safety and Handling

Safety and Handling

RTECSNumber : FF9335000

TSCA : TSCA

Documents