Please login to your online account to display your discounted pricing

Calcium carbonate, 97%, pure, chunks, ACROS Organics™


Quantity 3kg
Packaging Plastic bottle

Manufacturer: fisher scientific acros  403790030

 View more versions of this product

Catalog No. AC403790030

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 100.09
Physical Form Chunks
Quantity 3kg
Color White-beige to slightly beige-gray
Merck Index 15, 1659
TSCA TSCA
RTECS Number FF9335000
EINECS Number 207-439-9
Purity Grade Pure
Fieser 01,103; 02,57; 04,67; 05,89
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 97%
CAS 471-34-1
Molecular Formula CCaO3
Chemical Name or Material Calcium carbonate
MDL Number MFCD00010906
Linear Formula CaCO3
Name Note 97%
Packaging Plastic bottle
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 1 HYGROSCOPIC

Specifications

Chemical Name or Material Calcium carbonate
Name Note 97%
CAS 471-34-1
Purity Grade Pure
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula CCaO3
Linear Formula CaCO3
Formula Weight 100.09
MDL Number MFCD00010906
Accessories

HYGROSCOPIC

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents