missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium carbonate, 97%, pure, chunks, ACROS Organics™

$103.81 - $136.11

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
Synonym aeromatt
PubChem CID 10112
ChEBI CHEBI:3311
IUPAC Name calcium;carbonate
SMILES C(=O)([O-])[O-].[Ca+2]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC403790020 SDS View Documents Thermo Fisher Scientific
403790020
2kg Plastic bottle Each for $103.81
 
AC403790030 SDS View Documents Fisher Scientific Acros
403790030
3kg Plastic bottle Each for $136.11
 
Specifications

Chemical Identifiers

471-34-1
100.09
VTYYLEPIZMXCLO-UHFFFAOYSA-L
10112
calcium;carbonate
CCaO3
MFCD00010906
aeromatt
CHEBI:3311
C(=O)([O-])[O-].[Ca+2]

Specifications

471-34-1
CCaO3
MFCD00010906
15, 1659
Solubility in water: insoluble. Other solubilities: soluble in diluted acids
C(=O)([O-])[O-].[Ca+2]
100.09
CHEBI:3311
Chunks
Pure
Solubility in water: insoluble. Other solubilities: soluble in diluted acids
825.0°C
Calcium carbonate, 97%
96% min. (Complexometry)
CaCO3
01,103; 02,57; 04,67; 05,89
aeromatt
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium;carbonate
10112
100.09
97%
Plastic bottle
Beige-Gray to Beige-White
2kg
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

HYGROSCOPIC

EINECSNumber : 207-439-9

RTECSNumber : FF9335000

TSCA : TSCA