Please login to your online account to display your discounted pricing

Calcium Carbonate

$42.95 - $42.95

Chemical Identifiers

Chemical Name or Material Calcium Carbonate
CAS 471-34-1
Assay Percent Range 100% w/w
Linear Formula CaCO3
Formula Weight 100.09
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

S25220C

  SDS
aqua phoenix scientific
S25220C
Reagent Grade - Solid 2.5kg Each for $42.95
Description & Specifications

Specifications

Chemical Name or Material Calcium Carbonate
Shelf Life 5 years
CAS 471-34-1
ChemAlert Storage Symbol Gray
Assay Percent Range 100% w/w
Linear Formula CaCO3
Formula Weight 100.09
Density ∽2.7 to 2.9g/mL
Packaging Reagent Grade - Solid
Color White
Quantity 2.5kg

CO2 generation

This product is an Educational Grade Chemical — no C of A is available.

Safety and Handling
 

5 years