Learn More
Calcium carbide, 72-82%, Thermo Scientific Chemicals
$54.75 - $111.10
Chemical Identifiers
CAS | 75-20-7 |
---|---|
Molecular Formula | C2Ca |
Molecular Weight (g/mol) | 64.1 |
MDL Number | MFCD00010905 |
Synonym | calcium carbide, 3-phenoxypropylene di acetate, glycerol phenyl ether diacetate, 3-phenoxy-1,2-propanediol diacetate, phenylglyceryl ether diacetate, 1-phenyl ether-2,3-diacetate glycerol, 1,2-propanediol, 3-phenoxy-, diacetate, 1-acetyloxy-3-phenoxypropan-2-yl acetate, quartz crystal, 3-phenoxypropane-1,2-diyl diacetate |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC389792500
|
Thermo Scientific Chemicals
389792500 |
250 g | Glass bottle |
N/A
|
|
||||
AC389790010
|
Thermo Scientific Chemicals
389790010 |
1 kg | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75-20-7 | |
64.1 | |
calcium carbide, 3-phenoxypropylene di acetate, glycerol phenyl ether diacetate, 3-phenoxy-1,2-propanediol diacetate, phenylglyceryl ether diacetate, 1-phenyl ether-2,3-diacetate glycerol, 1,2-propanediol, 3-phenoxy-, diacetate, 1-acetyloxy-3-phenoxypropan-2-yl acetate, quartz crystal, 3-phenoxypropane-1,2-diyl diacetate |
C2Ca | |
MFCD00010905 |
Specifications
2300.0°C | |
Gray | |
Lump | |
12 to 23% (CaO) (Complexometry) | |
CaC2 | |
15, 1658 | |
Solubility in water: hydrolyses | |
64.1 | |
Positive | |
Calcium carbide |
2160.0°C | |
75-20-7 | |
250 g | |
C2Ca | |
MFCD00010905 | |
calcium carbide, 3-phenoxypropylene di acetate, glycerol phenyl ether diacetate, 3-phenoxy-1,2-propanediol diacetate, phenylglyceryl ether diacetate, 1-phenyl ether-2,3-diacetate glycerol, 1,2-propanediol, 3-phenoxy-, diacetate, 1-acetyloxy-3-phenoxypropan-2-yl acetate, quartz crystal, 3-phenoxypropane-1,2-diyl diacetate | |
64.1 | |
72 to 82% | |
Glass bottle |
Safety and Handling
GHS H Statement
In contact with water releases flammable gases which may ignite spontaneously.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye damage.
GHS P Statement
Keep away from any possible contact with water,because of violent reaction and possible flash fire.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protecti
GHS Signal Word: Danger
EINECSNumber : 200-848-3
RUO – Research Use Only