Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium bromide, ultra dry , 99.978% (metals basis)

Chemical Identifiers

Chemical Name or Material Calcium bromide
CAS 7789-41-5
Name Note Ultra dry
Linear Formula CaBr2
Formula Weight 199.9
MDL Number MFCD00010902
Merck Index 14,1654
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3568406

  SDS
alfa aesar™
3568406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3568403

  SDS
alfa aesar™
3568403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 199.9
Packaging Ampule
Chemical Name or Material Calcium bromide
TSCA TSCA
Mesh Size −10 Mesh
Odor Odorless
Name Note Ultra dry
Physical Form Beads
Assay 99.978%
Sensitivity Hygroscopic
Assay Percent Notes (metals basis)
Linear Formula CaBr2
MDL Number MFCD00010902
CAS 7789-41-5
Merck Index 14,1654
Boiling Point 806° to 812°C
Color White to off-white
Melting Point 730°C
Quantity 5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1
  • Ampouled under argon
Density 3.353g/mL
RTECS Number EV9328000

  • Ampouled under argon

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

  • Ampouled under argon