missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Calcium bromide hydrate, for analysis, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 300845000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Calcium bromide hydrate | |
For analysis | |
71626-99-8 | |
500 g | |
CaBr2·xH2O | |
Solubility in water: 1420g/L (20°C). Other solubilities: soluble in methanol, ethanol | |
[Ca++].[Br-].[Br-] | |
199.89 | |
≥83.0% (Complexometry) (CaBr2) | |
Plastic bottle |
730.0°C | |
White | |
Crystalline Powder and/or Chunks | |
Br2Ca | |
MFCD00149608 | |
WGEFECGEFUFIQW-UHFFFAOYSA-L | |
calcium dibromide | |
199.9 | |
Analytical |
Chemical Identifiers
71626-99-8 | |
199.89 | |
WGEFECGEFUFIQW-UHFFFAOYSA-L | |
[Ca++].[Br-].[Br-] |
Br2Ca | |
MFCD00149608 | |
calcium dibromide |
Safety and Handling
GHS H Statement
Causes serious eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Danger
RUO – Research Use Only