missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium bromide hydrate, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 4449736

 View more versions of this product

Catalog No. AA4449736



Specifications

Specifications

Calcium bromide hydrate
(metals basis)
MFCD00149608
Crystalline
White
71626-99-8
CaBr2·xH2O
199.90 (Anhydrous)
99.999%
500g

Chemical Identifiers

71626-99-8
CaBr2·xH2O
MFCD00149608
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P280-P305+P351+P338-P310

H318

EINECSNumber : 232-164-6

RTECSNumber : EV9328000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents