missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium bromide, anhydrous, 99.5%, Alfa Aesar™

For Photography, Dehydrating Agent, Manufacturing of Mineral Water

Manufacturer:  Alfa Aesar 8789318

 View more versions of this product

Catalog No. AA8789318



Specifications

Specifications

Calcium bromide
7789-41-5
Br2Ca
White to off-white powder or beads
WGEFECGEFUFIQW-UHFFFAOYSA-L
calcium;dibromide
5360316
199.9
White
Anhydrous
99.5%
MFCD00010902
calcium bromide, calcium dibromide, kalziumbromid, brocal, 87cny2eebh, ksc377c0p, calciumbromide, calcium 2+ dibromide, calcium bromide, anhydrous 10g
[Ca+2].[Br-].[Br-]
199.886
CHEBI:31338
Powder
50g

Chemical Identifiers

7789-41-5
199.886
WGEFECGEFUFIQW-UHFFFAOYSA-L
5360316
calcium;dibromide
Br2Ca
MFCD00010902
calcium bromide, calcium dibromide, kalziumbromid, brocal, 87cny2eebh, ksc377c0p, calciumbromide, calcium 2+ dibromide, calcium bromide, anhydrous 10g
CHEBI:31338
[Ca+2].[Br-].[Br-]
Safety and Handling

Safety and Handling

H315-H319-H335 : Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

EINECSNumber : 232-164-6

SDS Documents