Learn More
Calcium bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals
$70.01 - $1043.16
Chemical Identifiers
CAS | 165324-09-4 |
---|---|
Molecular Formula | C4CaF12N2O8S4 |
Molecular Weight (g/mol) | 600.349 |
MDL Number | MFCD14156019 |
InChI Key | WFABOCFDABTAPE-UHFFFAOYSA-N |
Synonym | calcium bis trifluoromethylsulfonyl imide, calcium ii bis trifluoromethanesulfonyl imide, calcium ii triflimide, calcium trifluoromethanesulfonimide, calcium bistrifluoromethanesulfonimide, calcium bis trifluoromethylsulfonyl amide, calcium bis bistriflylimide anion, calcium bis trifluoromethyl sulfonyl amide, bis bis trifluoromethylsulfonyl amino calcium, bis trifluoromethanesulfonyl imide calcium ii salt |
PubChem CID | 46242197 |
IUPAC Name | calcium;bis(trifluoromethylsulfonyl)azanide |
SMILES | C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Ca+2] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA46533MD
|
Thermo Scientific Chemicals
046533MD |
250 mg |
Each for $70.01
|
|
|||||
AA4653303
|
Thermo Scientific Chemicals
04653303 |
1 g |
Each for $235.16
|
|
|||||
AA4653306
|
Thermo Scientific Chemicals
04653306 |
5 g |
Each for $1,043.16
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
165324-09-4 | |
600.349 | |
WFABOCFDABTAPE-UHFFFAOYSA-N | |
46242197 | |
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Ca+2] |
C4CaF12N2O8S4 | |
MFCD14156019 | |
calcium bis trifluoromethylsulfonyl imide, calcium ii bis trifluoromethanesulfonyl imide, calcium ii triflimide, calcium trifluoromethanesulfonimide, calcium bistrifluoromethanesulfonimide, calcium bis trifluoromethylsulfonyl amide, calcium bis bistriflylimide anion, calcium bis trifluoromethyl sulfonyl amide, bis bis trifluoromethylsulfonyl amino calcium, bis trifluoromethanesulfonyl imide calcium ii salt | |
calcium;bis(trifluoromethylsulfonyl)azanide |
Specifications
165324-09-4 | |
C4CaF12N2O8S4 | |
MFCD14156019 | |
UN1759 | |
calcium bis trifluoromethylsulfonyl imide, calcium ii bis trifluoromethanesulfonyl imide, calcium ii triflimide, calcium trifluoromethanesulfonimide, calcium bistrifluoromethanesulfonimide, calcium bis trifluoromethylsulfonyl amide, calcium bis bistriflylimide anion, calcium bis trifluoromethyl sulfonyl amide, bis bis trifluoromethylsulfonyl amino calcium, bis trifluoromethanesulfonyl imide calcium ii salt | |
WFABOCFDABTAPE-UHFFFAOYSA-N | |
calcium;bis(trifluoromethylsulfonyl)azanide | |
46242197 | |
Solid |
>300°C | |
[(CF3SO2)2N]2Ca | |
250 mg | |
Moisture sensitive | |
Insoluble in water. | |
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Ca+2] | |
600.349 | |
600.35 | |
Calcium bis (trifluoromethylsulfonyl)imide |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only