missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium benzoate hydrate, 98%, water <8%

Manufacturer:  Alfa Aesar™ A123580E

Catalog No. AAA123580E



Description

Description

Applications
Calcium benzoate hydrate is used in used in the food industry as a preservative.

Solubility
Soluble in water (2.6 g/100ml at 0°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

Calcium benzoate hydrate
2090-05-3
C14H10CaO4
Benzoic acid calcium salt
C1=CC=C(C=C1)C(=O)[O-].C1=CC=C(C=C1)C(=O)[O-].[Ca+2]
282.31
282.32 (Anhydrous)
Water <8%
98%
MFCD00167164
NSQPPSOSXWOZNH-UHFFFAOYSA-L
calcium;dibenzoate
62425
2500g
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents