Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium alpha-D-isosaccharinate, 98% (Assay)

Chemical Identifiers

Chemical Name or Material Calcium alpha-D-isosaccharinate
Molecular Formula C12H22CaO12
Linear Formula [CO2C(OH)(CH2OH)CH2HCOHCH2OH]2Ca
Formula Weight 398.38
MDL Number MFCD04039767
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4437101

  SDS
alfa aesar™
4437101
0.25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4437114

  SDS
alfa aesar™
4437114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4437103

  SDS
alfa aesar™
4437103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4437106

  SDS
alfa aesar™
4437106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Calcium alpha-D-isosaccharinate
Physical Form Crystalline
Assay 98%
Assay Percent Notes (Assay)
Molecular Formula C12H22CaO12
Linear Formula [CO2C(OH)(CH2OH)CH2HCOHCH2OH]2Ca
Formula Weight 398.38
MDL Number MFCD04039767
Color Pale brown
Quantity 0.25g