missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Calcium alpha-D-isosaccharinate, 98% (Assay)

Manufacturer:  Alfa Aesar™ 4437103

 View more versions of this product

Catalog No. AA4437103

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Calcium alpha-D-isosaccharinate
(Assay)
[CO2C(OH)(CH2OH)CH2HCOHCH2OH]2Ca
398.38
98%
1g
84324-95-8
C12H22CaO12
MFCD04039767
Crystalline
Brown

Chemical Identifiers

84324-95-8
C12H22CaO12
MFCD04039767
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents