missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium acetate monohydrate, White powder, 99%, Alfa Aesar™

Manufacturer:  Alfa Aesar 1344630

 View more versions of this product

Catalog No. AA1344630



Specifications

Specifications

Calcium acetate monohydrate
99%
MFCD00150019
XQKKWWCELHKGKB-UHFFFAOYSA-L
calcium;diacetate;hydrate
82163
176.18
ACS
White
5743-26-0
C4H8CaO5
calcium acetate monohydrate, calcium diacetate monohydrate, unii-7za48gim5h, calcium diacetate hydrate, acetic acid, calcium salt, monohydrate, calcium acetate, monohydrate, 7za48gim5h, calcium acetat, calcium acetate jan, acmc-1c5pu
CC(=O)[O-].CC(=O)[O-].O.[Ca+2]
176.181
CHEBI:59199
Powder
1.5g/cm3
250g

Chemical Identifiers

5743-26-0
176.181
XQKKWWCELHKGKB-UHFFFAOYSA-L
82163
calcium;diacetate;hydrate
C4H8CaO5
MFCD00150019
calcium acetate monohydrate, calcium diacetate monohydrate, unii-7za48gim5h, calcium diacetate hydrate, acetic acid, calcium salt, monohydrate, calcium acetate, monohydrate, 7za48gim5h, calcium acetat, calcium acetate jan, acmc-1c5pu
CHEBI:59199
CC(=O)[O-].CC(=O)[O-].O.[Ca+2]
Safety and Handling

Safety and Handling

H303: May be harmful if swallowed.

EINECSNumber : 200-540-9

Documents